Tamper evident closures for VEREL bottles
Catalog # MUEL14.061.01
Supplier: MUELLER KREMPEL
Specifications
- Farbe:Weiß
- Material:PP
- VE:100
Specifications
About this item
Closure, tamper evident, with yellow safety ring and PE- foamed insert.
- Used for VERAL bottles of glass and PET