Anti-DRAM2 Rabbit Polyclonal Antibody
BIRBORB183201-100
: Biorbyt
- Antikörper-Typ:Primär
- Antigen-Symbol:DRAM2
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Lagertemperatur:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at –20 °C, avoid freeze/thaw cycles
- Immunogen:KLH conjugated synthetic peptide derived from human DRAM2
- VE:100 µG
Type: Primary
Antigen: DRAM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat