Anti-PTPRC Mouse Monoclonal Antibody [clone: UCHL-1+T200/797]
ABNOMAB14693
: Abnova
- Antikörper-Typ:Primär
- Antigen-Name:Cultured human T-cells from an IL-2-dependent T-cell line (CA1) (UCHL-1) and Human T-lymphocytes (T200/797)
- Antigen-Symbol:PTPRC
- Klonalität:Monoklonal
- Klon:UCHL-1+T200/797
- Wirt:Mouse
- ImmunoChemie:Yes
- Isotyp:IgG2a kappa
- Reaktivität:Human
- Form:Liquid
- Gen-ID:5788
- Lagerungspuffer:In 10 mM PBS (0.05% BSA and 0.05% azide).
- Lagertemperatur:Store at 4°C.
- Immunogen:Cultured human T-cells from an IL-2-dependent T-cell line (CA1) (UCHL-1) and Human T-lymphocytes (T200/797).
- Reinigung:Protein A/G purification
- Größe:100 µg
- VE:100 µG
Mouse monoclonal antibody raised against human CD45RO.
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: UCHL-1+T200/797
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human