Order Entry
Austria
ContactUsLinkComponent
 
Anti-ELMO1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-5807R-A647
 :  Bioss
Anti-ELMO1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-5807R-A647
 :  Bioss

 

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    engulfment and cell motility 1
  • Antigen-Symbol:
    ELMO1
  • Klonalität:
    Polyklonal
  • Konjugation:
    Alexa Fluor® 647
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Kreuzadsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    9844
  • Antigen-Synonyme:
    MGC126406|ced-12 homolog 1|ELMO-1|engulfment and cell motility 1|KIAA0281|CED12|CED-12
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagertemperatur:
    Store at 4°C for 12 months
  • Konzentration:
    1 μg/μl
  • Transporttemperatur:
    4°C
  • Reinigung:
    Purified by Protein A
  • VE:
    100 µl

 

 

ELMO1 is involved in cytoskeletal rearrangements required for phagocytosis of apoptotic cells and cell motility. ELMO1 acts in association with DOCK1 and CRK and was initially proposed to be required in complex with DOCK1 to activate Rac Rho small GTPases. It is belived to enhance the guanine nucleotide exchange factor (GEF) activity of DOCK1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ELMO1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat