Order Entry
Austria
ContactUsLinkComponent
Anti-ICAM1 Rabbit Polyclonal Antibody (Cy7®)
Artikelnr. BOSSBS-4618R-CY7
Lieferant:  Bioss
Anti-ICAM1 Rabbit Polyclonal Antibody (Cy7®)
Artikelnr. BOSSBS-4618R-CY7
Lieferant:  Bioss

Spezifikationen

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Intercellular Adhesion molecule 1
  • Antigen-Symbol:
    ICAM1
  • Klonalität:
    Polyklonal
  • Konjugation:
    Cy7®
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human
  • Kreuzadsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    3383
  • Antigen-Synonyme:
    ICAM-1|major group rhinovirus receptor|human rhinovirus receptor|BB2|cell surface glycoprotein P3.58|intercellular adhesion molecule 1|P3.58
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagertemperatur:
    Store at 4°C for 12 months
  • Konzentration:
    1 μg/μl
  • Transporttemperatur:
    4°C
  • Immunogen:
    45-95/532
  • Reinigung:
    Purified by Protein A
  • VE:
    100 µl

Spezifikationen

Über diesen Artikel

ICAM proteins are ligands for the leukocyte adhesion protein LFA-1 (integrin alpha-L/beta-2). During leukocyte trans-endothelial migration, ICAM1 engagement promotes the assembly of endothelial apical cups through ARHGEF26/SGEF and RHOG activation. In case of rhinovirus infection acts as a cellular receptor for the virus.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ICAM1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human