Order Entry
Austria
ContactUsLinkComponent
Anti-RGS5 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
  BOSSBS-2794R-A488
 :  Bioss
Anti-RGS5 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
  BOSSBS-2794R-A488
 :  Bioss

 

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Regulator of G-protein Signaling 5
  • Antigen-Symbol:
    RGS5
  • Klonalität:
    Polyklonal
  • Konjugation:
    Alexa Fluor® 488
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Rat,
    Mouse
  • Kreuzadsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    19737
  • Antigen-Synonyme:
    MSTP092|MSTP032|MSTP129|MST106|MSTP106|MST092|MST129
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagertemperatur:
    Store at 4°C for 12 months
  • Konzentration:
    1 μg/μl
  • Transporttemperatur:
    4°C
  • Immunogen:
    80-130/181
  • Reinigung:
    Purified by Protein A
  • VE:
    100 µl

 

 

Inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. Binds to G(i)-alpha and G(o)-alpha, but not to G(s)-alpha.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RGS5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat