Order Entry
Austria
ContactUsLinkComponent
Anti-TGFB5/ TGFB1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Artikelnr. BOSSBS-3846R-A647
Lieferant:  Bioss
Anti-TGFB5/ TGFB1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Artikelnr. BOSSBS-3846R-A647
Lieferant:  Bioss

Spezifikationen

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Transforming Growth Factor beta 5/ Transforming Growth Factor beta 1
  • Antigen-Symbol:
    TGFB5/ TGFB1
  • Klonalität:
    Polyklonal
  • Konjugation:
    Alexa Fluor® 647
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Multiple
  • Kreuzadsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    397778
  • Antigen-Synonyme:
    Transforming growth factor beta 5|TGF-beta-1|TGF beta 5|Transforming growth factor beta-1.TGFB1_XENLA|Transforming Growth Factor 5|TGF-beta-5
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagertemperatur:
    Store at 4°C for 12 months
  • Konzentration:
    1 μg/μl
  • Transporttemperatur:
    4°C
  • Reinigung:
    Purified by Protein A
  • VE:
    100 µl

Spezifikationen

Über diesen Artikel

Transforming Growth Factor beta 5 (TGF beta 5) is a member of the TGF beta family of growth factors. The TGF beta polypeptides are multifunctional; capable of influencing cell proliferation, differentiation, and other functions in a wide range of cell types. Transformed, as well as nonneoplastic tissues, release transforming growth factors; and essentially all mammalian cells possess a specific TGF receptor. The multi-modal nature of TGF beta is seen in its ability to stimulate or inhibit cellular proliferation. In general, cells of mesenchymal origin appear to be stimulated by TGF beta whereas cells of epithelial or neuroectodermal origin are inhibited by the peptide.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TGF beta 5/ TGF beta 1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species