Order Entry
Austria
ContactUsLinkComponent
 
Anti-EIF4E Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-2446R-FITC
 :  Bioss
Anti-EIF4E Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-2446R-FITC
 :  Bioss

 

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    eukaryotic translation initiation factor 4E
  • Antigen-Symbol:
    EIF4E
  • Klonalität:
    Polyklonal
  • Konjugation:
    FITC (Fluorescein Isothiocyanate)
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Kreuzadsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    1977
  • Antigen-Synonyme:
    eIF-4F 25 kDa subunit|EIF4EL1|EIF4E1|eukaryotic translation initiation factor 4E-like 1|eIF-4E|CBP|eIF4E|EIF4F|mRNA cap-binding protein
  • Modification:
    phospho-Ser209
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagertemperatur:
    Store at 4°C for 12 months
  • Konzentration:
    1 μg/μl
  • Transporttemperatur:
    4°C
  • Immunogen:
    200-217/217
  • Reinigung:
    Purified by Protein A
  • VE:
    100 µl

 

 

Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures. Component of the CYFIP1-EIF4E-FMR1 complex which binds to the mRNA cap and mediates translational repression. In the CYFIP1-EIF4E-FMR1 complex this subunit mediates the binding to the mRNA cap.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EIF4E
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat