Order Entry
Austria
ContactUsLinkComponent
Anti-MITF Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-1990R-CY5
Supplier:  Bioss
Anti-MITF Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-1990R-CY5
Supplier:  Bioss

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Microphthalmia-Associated Transcription Factor
  • Antigen-Symbol:
    MITF
  • Klonalität:
    Polyklonal
  • Konjugation:
    Cy5®
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Kreuzadsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    4286
  • Antigen-Synonyme:
    bHLHe32|BHLHE32|MITF|Class E basic helix-loop-helix protein 32|Microphthalmia-associated transcription factor
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagertemperatur:
    Store at 4°C for 12 months
  • Konzentration:
    1 μg/μl
  • Transporttemperatur:
    4°C
  • Reinigung:
    Purified by Protein A
  • VE:
    100 µl

Specifications

About this item

Transcription factor that regulates the expression of genes with essential roles in cell differentiation, proliferation and survival. Binds to symmetrical DNA sequences (E-boxes) (5'-CACGTG-3') found in the promoters of target genes, such as BCL2 and tyrosinase (TYR). Plays an important role in melanocyte development by regulating the expression of tyrosinase (TYR) and tyrosinase-related protein 1 (TYRP1). Plays a critical role in the differentiation of various cell types, such as neural crest-derived melanocytes, mast cells, osteoclasts and optic cup-derived retinal pigment epithelium.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MITF
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat