Order Entry
Austria
ContactUsLinkComponent
Anti-PCD5 Rabbit Polyclonal Antibody (Cy5®)
Artikelnr. BOSSBS-1333R-CY5
Lieferant:  Bioss
Anti-PCD5 Rabbit Polyclonal Antibody (Cy5®)
Artikelnr. BOSSBS-1333R-CY5
Lieferant:  Bioss

Spezifikationen

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Programmed Cell Death 5
  • Antigen-Symbol:
    PCD5
  • Klonalität:
    Polyklonal
  • Konjugation:
    Cy5®
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Kreuzadsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    9141
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagertemperatur:
    Store at 4°C for 12 months
  • Konzentration:
    1 μg/μl
  • Transporttemperatur:
    4°C
  • Reinigung:
    Purified by Protein A
  • VE:
    100 µl

Spezifikationen

Über diesen Artikel

This gene encodes a protein expressed in tumor cells during apoptosis independent of the apoptosis-inducing stimuli. Prior to apoptosis induction, this gene product is distributed in both the nucleus and cytoplasm. Once apoptosis is induced, the level of this protein increases and by relocation from the cytoplasm, it accumulates in the nucleus. Although its exact function is not defined, this protein is thought to play an early and universal role in apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TFAR19/PDCD5
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat