Reversed phase chromatography columns, RSpak NN
: Resonac
RSpak NN columns are ideal for analysis of complex samples containing neutral and ionic substances by multimode (reversed-phase and cation exchange) analysis.
- Packing material contains sulpho groups
- Base material is polyhydroxymethacrylate
- Catalog No:
- 554-2923
- 554-2885
- Bezeichnung:
- RSpak NN-814
- RSpak NN-G
- Max. Druck:
- 2,0 MPa (20 bar)
- pH-Bereich:
- 2 - 12
- 2 - 12
- Temperaturlimit:
- 10 - 60 °C
- 10 - 50 °C
Frequently Bought Together