Dehydrated culture media, potato dextrose agar, MP Biomedicals
Artikelnr. ICNA091008517
Lieferant: MP Biomedicals
CAS-Nummer:
Spezifikationen
- VE:500 g
- Normen:APHA
- Bezeichnung:Potato dextrose agar, powder
Spezifikationen
Über diesen Artikel
Potato dextrose agar (PDA) is suited for the cultivation of yeasts and moulds from dairy products and other foods. It may also be used to induce sporulation in many fungi.
Certifications: This medium is recommended by the Association of Public Health (APHA).