Total hardness reagent set, for digital titrator
HACH2272000
: Hach
:
- Chemische Methode:EDTA
- VE:100 Tests
- Bezeichnung:Hardness reagent set
- Messbereich:10 - 160 mg/l; 100 - 4000 mg/l (CaCO₃)
For total hardness determination by EDTA digital titrator titration with calmagite indicator.
- Accurate results
- Fast and flexible
- Versatile and economical
Digital titrator methods use a high-precision dispensation device and cartridges of concentrated titrant to allow analyses in the lab, the plant or the field. The interchangeable cartridges allow to change titrants in seconds without needing to clean or rinse burettes.
: Set contains ManVer® 2 indicator, hardness 1 buffer solution and one cartridge each of 0,0800 M and 0,800 M EDTA.