Alkaline detergents, powder, LaboClean F, neodisher®
Artikelnr. 148-3139
Lieferant: Dr. Weigert
Spezifikationen
- Bezeichnung:LaboClean F, Alkaline detergent
- VE:4
Spezifikationen
Über diesen Artikel
LaboClean F detergent is composed of alkaline components and dispersants. Ideal for the removal of grease, oil, paraffin, resin, organic dyes and residues of indelible pen.
- High complexing and dispersive action
- Used for intensive cleaning
- Free of phosphates and surfactants
It is not suitable for non ferrous and light metals.