Alkaline detergents, liquid concentrate, LaboClean FM, neodisher®
Catalog # 148-0159
Supplier: Dr. Weigert
Specifications
- Bezeichnung:LaboClean FM, Alkaline detergent
- VE:5 L
Specifications
About this item
LaboClean FM is an alkaline, surfactant-containing detergent with intensive cleaning action. Ideal for the removal of oil, fat, resin, paraffin and organic dyes.
- High emulsifying and dispersion action
- High soil carrying ability
- Free of phosphates, silicates and oxidising agents
Suitable for alkali-compatible utensils made of glass, stainless steel, ceramic and plastics. It is not suitable for utensils made of aluminium and nonanodised aluminium.