ContactUsLinkComponent
ValvTool wrench
ValvTool wrench
Catalog # REST25321
Supplier:  Restek
undefined
ValvTool wrench
Catalog # REST25321
Supplier:  Restek
Specifications
  • Bezeichnung:
    ValvTool wrench, for 1/4" nuts
  • Seq:
    01
  • VE:
    1
Specifications
About this item
The ValvTool is a time saving device that provides easy access to many hard-to-reach Rheodyne or Valco valves.