ValvTool wrench
Catalog # REST25321
Supplier: Restek
Specifications
- Bezeichnung:ValvTool wrench, for 1/4" nuts
- Seq:01
- VE:1
Specifications
About this item
The ValvTool is a time saving device that provides easy access to many hard-to-reach Rheodyne or Valco valves.