Anti-Na+ CP type IXalpha Rabbit Polyclonal Antibody
BWRLBS5813
- Antikörper-Typ:Primär
- Antigen-Symbol:Na+ CP type IX alpha
- Klonalität:Polyklonal
- Wirt:Rabbit
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
Synthetic peptide, corresponding to amino acids 702-747 of Human Na+ CP type IXalpha.
Na+ CP type IX? pAb detects endogenous levels of Na+ CP type IX? protein.
Type: Primary
Antigen: Na+ CP type IXalpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat