Order Entry
Austria
ContactUsLinkComponent
 
Anti-Na+ CP type IValpha Rabbit Polyclonal Antibody
  BWRLBS5812
Anti-Na+ CP type IValpha Rabbit Polyclonal Antibody
  BWRLBS5812

 

  • Antikörper-Typ:
    Primär
  • Antigen-Symbol:
    Na+ CP type IV alpha
  • Klonalität:
    Polyklonal
  • Wirt:
    Rabbit
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Größe:
    100 µg
  • VE:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 472-517 of Human Na+ CP type IValpha.

Na+ CP type IV? pAb detects endogenous levels of Na+ CP type IV? protein.

Type: Primary
Antigen: Na+ CP type IValpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat