Anti-p-Gab1 Rabbit Polyclonal Antibody [clone: Y659]
BWRLBS4689
- Antikörper-Typ:Primär
- Antigen-Symbol:p-Gab1
- Klonalität:Polyklonal
- Klon:Y659
- Wirt:Rabbit
- ImmunoChemie:Yes
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
Synthetic phosphopeptide derived from human Gab 1 around the phosphorylation site of Tyrosine 659.
p-Gab1 (Y659) pAb detects endogenous levels of Gab1 protein only when phosphorylated at Tyr659.
Type: Primary
Antigen: p-Gab1
Clonality: Polyclonal
Clone: Y659
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat