Anti-p-CXCR4 Rabbit Polyclonal Antibody [clone: S339]
Catalog # BWRLBS4688
Supplier: Bioworld Technology
Specifications
- Antikörper-Typ:Primär
- Antigen-Symbol:p-CXCR4
- Klonalität:Polyklonal
- Klon:S339
- Wirt:Rabbit
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human CXCR4 around the phosphorylation site of Serine 339.
p-CXCR4 (S339) pAb detects endogenous levels of CXCR4 protein only when phosphorylated at Ser339.
Type: Primary
Antigen: p-CXCR4
Clonality: Polyclonal
Clone: S339
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat