Anti-p-E2F1 Rabbit Polyclonal Antibody [clone: T433]
Artikelnr. BWRLBS4679
Supplier: Bioworld Technology
Specifications
- Antikörper-Typ:Primär
- Antigen-Symbol:p-E2F1
- Klonalität:Polyklonal
- Klon:T433
- Wirt:Rabbit
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human E2F1 around the phosphorylation site of Threonine 433.
p-E2F1 (T433) pAb detects endogenous levels of E2F1 protein only when phosphorylated at Thr433.
Type: Primary
Antigen: p-E2F1
Clonality: Polyclonal
Clone: T433
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat