Anti-p-NPM Rabbit Polyclonal Antibody [clone: T234]
Catalog # BWRLBS4656
Supplier: Bioworld Technology
Specifications
- Antikörper-Typ:Primär
- Antigen-Symbol:p-NPM
- Klonalität:Polyklonal
- Klon:T234
- Wirt:Rabbit
- ImmunoChemie:Yes
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human NPM around the phosphorylation site of Threonine 234.
p-NPM (T234) pAb detects endogenous levels of NPM protein only when phosphorylated at Thr234.
Type: Primary
Antigen: p-NPM
Clonality: Polyclonal
Clone: T234
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat