Order Entry
Austria
ContactUsLinkComponent
 
Anti-p-APC Rabbit Polyclonal Antibody [clone: S2054]
  BWRLBS4649
Anti-p-APC Rabbit Polyclonal Antibody [clone: S2054]
  BWRLBS4649

 

  • Antikörper-Typ:
    Primär
  • Antigen-Symbol:
    p-APC
  • Klonalität:
    Polyklonal
  • Klon:
    S2054
  • Wirt:
    Rabbit
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Größe:
    100 µg
  • VE:
    100 µG

 

 

Synthetic phosphopeptide derived from human APC around the phosphorylation site of Serine 2054.

p-APC (S2054) pAb detects endogenous levels of APC protein when phosphorylated at Ser2054.

Type: Primary
Antigen: p-APC
Clonality: Polyclonal
Clone: S2054
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat