Anti-N4BP1 Rabbit Polyclonal Antibody [clone: Y415]
Catalog # BWRLBS3340
Supplier: Bioworld Technology
Specifications
- Antikörper-Typ:Primär
- Antigen-Symbol:N4BP1
- Klonalität:Polyklonal
- Klon:Y415
- Wirt:Rabbit
- ImmunoChemie:Yes
- Reaktivität:Human
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 391-440 of Human N4BP1.
N4BP1 (Y415) pAb detects endogenous levels of N4BP1 protein.
Type: Primary
Antigen: N4BP1
Clonality: Polyclonal
Clone: Y415
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human