Anti-Neuroglycan C Rabbit Polyclonal Antibody [clone: W248]
BWRLBS3833
- Antikörper-Typ:Primär
- Antigen-Symbol:Neuroglycan C
- Klonalität:Polyklonal
- Klon:W248
- Wirt:Rabbit
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
Synthetic peptide, corresponding to amino acids 221-270 of Human Neuroglycan C.
Neuroglycan C (W248) pAb detects endogenous levels of Neuroglycan C protein.
Type: Primary
Antigen: Neuroglycan C
Clonality: Polyclonal
Clone: W248
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat