Anti-Trk A Rabbit Polyclonal Antibody [clone: A785]
BWRLBS1430
- Antikörper-Typ:Primär
- Antigen-Symbol:Trk A
- Klonalität:Polyklonal
- Klon:A785
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Reaktivität:Human
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
Synthetic peptide, corresponding to amino acids 751-800 of Human Trk A.
Trk A (A785) pAb detects endogenous levels of Trk A protein.
Type: Primary
Antigen: Trk A
Clonality: Polyclonal
Clone: A785
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human