Anti-GCS-alpha-1 Rabbit Polyclonal Antibody [clone: D423]
BWRLBS3670
- Antikörper-Typ:Primär
- Antigen-Symbol:GCS-alpha-1
- Klonalität:Polyklonal
- Klon:D423
- Wirt:Rabbit
- ImmunoChemie:Yes
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
Synthetic peptide, corresponding to amino acids 400-450 of Human GCS-alpha-1.
GCS-?-1 (D423) pAb detects endogenous levels of GCS-?-1 protein.
Type: Primary
Antigen: GCS-alpha-1
Clonality: Polyclonal
Clone: D423
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat