Anti-E2F-1 Rabbit Polyclonal Antibody [clone: D426]
Catalog # BWRLBS1397
Supplier: Bioworld Technology
Specifications
- Antikörper-Typ:Primär
- Antigen-Symbol:E2F-1
- Klonalität:Polyklonal
- Klon:D426
- Wirt:Rabbit
- ImmunoChemie:Yes
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 400-450 of Human E2F-1.
E2F1 (D426) pAb detects endogenous levels of E2F1 protein.
Type: Primary
Antigen: E2F-1
Clonality: Polyclonal
Clone: D426
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat