Anti-Patched Rabbit Polyclonal Antibody [clone: A2]
BWRLBS1286
- Antikörper-Typ:Primär
- Antigen-Symbol:Patched
- Klonalität:Polyklonal
- Klon:A2
- Wirt:Rabbit
- ImmunoChemie:Yes
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
Synthetic peptide, corresponding to the N-terminal of Human Patched.
Patched (A2) pAb detects endogenous levels of Patched protein.
Type: Primary
Antigen: Patched
Clonality: Polyclonal
Clone: A2
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat