Non-ionic detergent, Kolliphor® HS 15
Kolliphor® HS 15, 5,0 - 7,0
Artikelnr. SIAL42966-1KG
Anbieter: Merck
Springe zu:
Product Details & Documents
Kolliphor HS 15 is a non-ionic detergent obtained by reacting 15 moles of ethylene oxide with 1 mole of 12-hydroxy stearic acid. It's a non-ionic solubiliser and emulsifier.
Spezifikationen
- Bezeichnung:Kolliphor® HS 15
- pH-Bereich:5,0 - 7,0