AuftragsverfolgungKontakt
Confidence solution, 5580 | Avantor
Confidence solution, 5580
Confidence solution 5580, for verifying pH, ORP, and conductivity values, 6 plastic bottles (1 pint each), 1 KIT
Artikelnr. YSIA005580
Anbieter:   YSI ENVIRONMENTAL
Confidence solution, 5580
Artikelnr. YSIA005580
Anbieter:  YSI ENVIRONMENTAL
CAS Nummer:  
Confidence solution, 5580
Confidence solution, 5580
Confidence solution 5580, for verifying pH, ORP, and conductivity values, 6 plastic bottles (1 pint each), 1 KIT
CAS Nummer:  
Preis
246,00 €
Einloggen für Verfügbarkeit
1 * 1 KIT
Product Details & Documents
This Confidence Solution® helps users to ensure probe performance of their pH, ORP, or conductivity probes.
This solution is designed for verifying measurements, not calibration. Simply immerse your multi-parameter probes in the solution and verify the pH, ORP, and conductivity values instantly. Ideal for quality checks during or after your study.
Expiration dates: Unopened=1 year, opened=3 months.
Delivery information: Case includes 6 plastic bottles (1 pint each).
Spezifikationen
Spezifikationen
  • VE:
    1 KIT
  • Bezeichnung:
    Confidence solution 5580, for verifying pH, ORP, and conductivity values, 6 plastic bottles (1 pint each)
Spezifikationen