ContactUsLinkComponent
Low pressure slip-on inlet filter
Low pressure slip-on inlet filter
Catalog # REST25008
Supplier:  Restek
undefined
Low pressure slip-on inlet filter
Catalog # REST25008
Supplier:  Restek
Specifications
  • Bezeichnung:
    Slip-on inlet filter
  • VE:
    1
Specifications
About this item
This slip-on inlet filter is made of 316 stainless steel tip with a tefzel collar seals to a corrosion-resistant 316 stainless steel filter element.
  • Easily attaches to the pump inlet line, without the use of wrenches
  • Cylindrical filter with standard 10 μm porosity
The universal ¹/₈" Ext.Ø tip accommodates standard PTFE tubing inner diameters. Fits Altex, ISCO, LDC, Varian, Waters, PerkinElmer, and other pumps.