Installation kits for packed columns
Catalog # REST21067
Supplier: Restek
Specifications
- Bezeichnung:Installation Kit for 2 mm ID columns; for valve applications, Kit contains 1× stainless steel nut 3,2 mm, 1× stainless steel Valco nut, 1× Vespel/graphite ferrule 3,2 mm, 1× stainless steel Valco ferrule 3,2 mm, 1× stainless steel front ferrule 3,2 mm, 1× stainless steel back ferrule 3,2 mm
- VE:1 KIT