GC ferrules for Shimadzu
Lieferant: Restek
Zertifikate
Über diesen Artikel
Graphite two-piece construction. For Shimadzu 17A, 2010, and 2014 GCs.
Packaging: Packaged on mandrel for easy handling.
Frequently Bought Together
Graphite two-piece construction. For Shimadzu 17A, 2010, and 2014 GCs.