AuftragsverfolgungKontakt
Passive air sampling kits
Supplier:   Restek
Passive air sampling kits
Supplier:  Restek
Certificates
Passive air sampling kits
Passive air sampling kits
6 Options Available Below
Product Details & Documents
This unique grab sampler minimises connections and leaks and houses a critical orifice in the tee fitting.
  • Different orifice sizes for sampling from four minutes to ten hours
  • Compression sample inlet allows easy connection
  • Canister valve connected to a sample port tubing
  • Stainless steel tee with orifice and vaccum gauge
  • Frit filter for insertion into a compression sample inlet
Specifically designed for soil gas sampling by allowing the connection of tubing coming from the soil gas sample port.
 : Canisters and replacement tees and orifices can be ordered separately.
Product Family Options
Product Information
  • Bezeichnung
    Luftstrom
    Material
    Availability
    Price
  • Air sampler with 0,635 mm orifice
    30 ml/min
    Edelstahl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Air sampler with 0,635 mm orifice
    30 ml/min
    Siltek® behandeltes Edelstahl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Replacement parts
Product Information
  • Bezeichnung
    Luftstrom
    Material
    Availability
    Price
  • Replacement frit filters, 2 µm
    Edelstahl
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 64,00 €
    64,00 €
    1 * 3 ST
       64,00 €
  • Replacement frit filters, 2 µm
    Siltek® behandeltes Edelstahl
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 106,00 €
    106,00 €
    1 * 3 ST
       106,00 €
  • Replacement tees with 0,0635 mm orifice
    30 ml/min
    Edelstahl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Replacement tees with 0,0635 mm orifice
    30 ml/min
    Siltek® behandeltes Edelstahl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Replacement parts
Product Information
  • Catalog #
    Bezeichnung
    Material
    Luftstrom
    Availability
    Price
  • Replacement frit filters, 2 µm
    Siltek® behandeltes Edelstahl
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 106,00 €
    106,00 €
    1 * 3 ST
       106,00 €
Product Information
  • Catalog #
    Bezeichnung
    Material
    Luftstrom
    Availability
    Price
  • Replacement frit filters, 2 µm
    Edelstahl
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 64,00 €
    64,00 €
    1 * 3 ST
       64,00 €
Product Information
  • Catalog #
    Bezeichnung
    Material
    Luftstrom
    Availability
    Price
  • Replacement tees with 0,0635 mm orifice
    Edelstahl
    30 ml/min
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 0,00 €
    0,00 €
    1 * 1 ST
       0,00 €
Product Information
  • Catalog #
    Bezeichnung
    Material
    Luftstrom
    Availability
    Price
  • Replacement tees with 0,0635 mm orifice
    Siltek® behandeltes Edelstahl
    30 ml/min
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 0,00 €
    0,00 €
    1 * 1 ST
       0,00 €