Make life easier (MLE) capillary tool kit for Bruker/Varian GCs
Catalog # REST22184
Supplier: Restek
Specifications
- Bezeichnung:MLE capillary tool kit for Bruker/Varian GCs
- VE:1 KIT
Specifications
About this item
MLE tool kits conveniently provide the tools that make it easier to install and maintain capillary columns.
Delivery: Kit includes 1/8", 3/16", 1/4" nylon brushes, 1/4", 3/8", 3/16" stainless steel wire tube brushes, stainless steel surface brush, 6 stainless steel jet reamers (0,25 to 0,65 mm OD), 1/4×5/16" open end wrench, 3/8×7/16" open end wrench, rubber-tipped slide-lock tweezers, scoring wafers, inlet liner removal tool, septum puller, mini wool puller/inserter tool, 4-inch tapered needle file, swivel head flashlight, mini hand drill set, 15 cm compact steel ruler, pocket magnifier, high temperature string (1 m), pipe cleaner (12-inch), cotton tip swabs (pack of 25), capillary installation gauge, injector wrench, septum nut removal tool, 7/16×1/2" open end wrench and 1/2×9/16" open end wrench.