Protease inhibitor cocktail, MP Biomedicals
ICNA0215883701
:
- VE:1 KIT
- Bezeichnung:Protease inhibitor cocktail
A broad spectrum protease inhibitor cocktail which covers both endopeptidases and exopeptidases, non-specific and specific. The cocktail consists of four major protease inhibitors such as AEBSF, Sodium EDTA, Leupeptin and Pepstain A.