Recombinant ubiquitin (from E. coli), rhodamine
Catalog # ENZOBMLSE7610025
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- VE:25 µG
- Konjugation:Rhodamin
- Protein/Peptid-Typ:Rekombinant
- Ursprung:E. coli
- Seq:02
- Gen-ID:P62988
- Protein/Peptid-Name:Ubiquitin
- Purity:>95% (HPLC)
- Molecular weight:~8,9 kDa
- Zusammensetzung:Liquid in dimethylsulphoxide
- Transporttemperatur:Dry Ice
Specifications
About this item
Ubiquitin-rhodamine is a high purity industry standard substrate for studying deubiquitinylating activity where detection sensitivity or continuous monitoring of activity.
- Molecular weight: ~8,9 kDa
It is prepared by the C-terminal derivatisation of ubiquitin with modified rhodamine-110 and has been shown to be a useful and sensitive fluorogenic substrate for wide range of deubiquitinylating enzymes (DUB's), including ubiquitin C-terminal hydrolases (UCH's) and ubiquitin specific proteases (USP's)