Human recombinant IL8
: Biorbyt
- Protein/Peptid-Typ:Rekombinant
- Spezies:Human
- Lagerbedingungen:Lyophilized Nafarelin although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution NAF should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Proteinsynonym:310C
- Protein/Peptid-Name:IL8
- Purity:Greater than 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized Nafarelin in sterile H2O not less than 100 ug/ml, which can then be further diluted to other aqueous solutions.
- Zusammensetzung:The protein (1 mg/ml) was lyophilized with no additives.
- Validierte Anwendungen:SDS-PAGE
Frequently Bought Together