Human CD4 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Ursprung:E. coli
- Spezies:Human
- Lagerbedingungen:Store vial at −20 °C to -80oC. When stored at the recommended temperature, this protein is stable for One year. Please avoid freeze-thaw cycles.
- Proteinsynonym:CD 4
- Protein/Peptid-Name:CD4
- Zusammensetzung:Each mg contains 1X PBS, 0.1% SDS and 1mM DTT.
- Validierte Anwendungen:SDS-PAGE , HPLC , FuncS