Human recombinant CEBP-g (from E. coli)
Lieferant: Biorbyt
Zertifikate
Über diesen Artikel
Spezifikationen
- Protein/Peptid-Typ:Rekombinant
- Ursprung:E. coli
- Spezies:Human
- Lagerbedingungen:Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA). Avoid multiple freeze-thaw cycles.
- Proteinsynonym:CCAAT
- Protein/Peptid-Name:CEBP-g
- Purity:Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Sequence:MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMPGG GGKAVAPSKQ SKKSSPMDRN SDEYRQRRER NNMAVKKSRL KSKQKAQDTL QRVNQLKEEN ERLEAKIKLL TKELSVLKDL FLEHAHNLAD NVQSISTENT TADGDN.
- Zusammensetzung:The protein contains 20 mM Tris-HCl pH7.5, 0.1M NaCl and 5mM b-Mercaptoethanol.
- Validierte Anwendungen:SDS-PAGE , HPLC , FuncS