Human BMP7 (from E. coli)
Lieferant: Biorbyt
Zertifikate
Über diesen Artikel
Spezifikationen
- Ursprung:E. coli
- Spezies:Human
- Lagerbedingungen:Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. Please avoid freeze thaw cycles.
- Proteinsynonym:BMP 7
- Protein/Peptid-Name:BMP7
- Purity:Greater than 95.0% as determined by SDS-PAGE.
- Sequence:MSTGSKQRSQ NRSKTPKNQE ALRMANVAEN SSSDQRQACK KHELYVSFRD LGWQDWIIAP EGYAAYYCEG ECAFPLNSYM NATNHAIVQTLVHFINPETV PKPCCAPTQL NAISVLYFDD SSNVILKKYR NMVVRACGCH LEHHHHHH.
- Zusammensetzung:BMP-7 protein solution contains 10mM sodium citrate pH=3.5 and 10% glycerol.
- Validierte Anwendungen:SDS-PAGE , HPLC , FuncS