Order Entry
Austria
ContactUsLinkComponent
Recombinant Influenza A, Avian, Vietnam 1203/04, H5N1, Hemagglutinin
Artikelnr. BIRBORB108507-10
Lieferant:  Biorbyt
CAS-Nummer:  
Recombinant Influenza A, Avian, Vietnam 1203/04, H5N1, Hemagglutinin
Artikelnr. BIRBORB108507-10
Lieferant:  Biorbyt
CAS-Nummer:  

Spezifikationen

  • VE:
    10 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Lagerbedingungen:
    Aliquot and store at −20 °C. Avoid repeated freeze/thaw cycles.
  • Protein/Peptid-Name:
    Influenza A, Avian, Vietnam 1203/04, H5N1, Hemagglutinin
  • Purity:
    Greater than 90% as determined by: Analysis by RP-HPLC, Anion-exchange FPLC or Analysis by reducing and non-reducing SDS-PAGE Silver Stained gel.

Spezifikationen

Über diesen Artikel