Order Entry
Austria
ContactUsLinkComponent
Recombinant HCV NS5a
Catalog # BIRBORB98871-1
Supplier:  Biorbyt
CAS Number:  
Recombinant HCV NS5a
Catalog # BIRBORB98871-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • VE:
    1 mg
  • Protein/Peptid-Typ:
    Rekombinant
  • Lagerbedingungen:
    Store at −20 °C. Avoid multiple freeze/thaw cycles.
  • Protein/Peptid-Name:
    HCV NS5a
  • Purity:
    Greater than 95% pure SDS PAGE non-reducing, Electrophoresis
  • Zusammensetzung:
    PBS, pH 7.4
  • Validierte Anwendungen:
    ELISA

Specifications

About this item