Order Entry
Austria
ContactUsLinkComponent
Rabbit Troponin Complex
Rabbit Troponin Complex
Catalog # BIRBORB79701-0.1
Supplier:  Biorbyt
CAS Number:  
Rabbit Troponin Complex
Catalog # BIRBORB79701-0.1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • VE:
    0,1 mg
  • Spezies:
    Rabbit
  • Lagerbedingungen:
    Store at -2 to -8 °C.
  • Protein/Peptid-Name:
    Troponin Complex
  • Purity:
    Greater than 95% pure SDS-PAGE. Protein G Chromatography

Specifications

About this item