Order Entry
Austria
ContactUsLinkComponent
Human recombinant SDF1 alpha (from E. coli)
Human recombinant SDF1 alpha (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant SDF1 alpha (from E. coli)
Supplier:  Biorbyt

Specifications

  • Konjugation:
    Unconjugated
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Human
  • Lagerbedingungen:
    Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Proteinsynonym:
    C-X-C motif chemokine 12
  • UniProtKB:
    P48061 (Human)
  • Protein/Peptid-Name:
    SDF1 alpha
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    KPVSLSYRCPCRFFESHVARANVKHLKILNTPNCALQIVARLKNNNRQVCIDPKLKWIQEYLEKA LNK
  • Zusammensetzung:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2.
  • Validierte Anwendungen:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together