ContactUsLinkComponent
Human recombinant Cathepsin S (from HEK293 cells)
Lieferant:  Biorbyt
Zertifikate
Human recombinant Cathepsin S (from HEK293 cells)
Lieferant:  Biorbyt
Spezifikationen
  • Konjugation:
    Unconjugated
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    HEK293 cells
  • Spezies:
    Human
  • Lagerbedingungen:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • UniProtKB:
    P25774 (Human)
  • Protein/Peptid-Name:
    Cathepsin S
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    QLHKDPTLDHHWHLWKKTYGKQYKEKNEEAVRRLIWEKNLKFVMLHNLEHSMGMHSYDLGMNHLG DMTSEEVMSLMSSLRVPSQWQRNITYKSNPNWILPDSVDWREKGCVTEVKYQGSCGACWAFSAVG ALEAQLKLKTGKLVSLSAQNLVDCSTEKYGNKGCNGGFMTTAFQYIIDNKGIDSDASYPYKAMDQ KCQYDSKYRAATCSKYTELPYGREDVLKEAVANKGPVSVGVDARHPSFFLYRSGVYYEPSCTQNV NHGVLVVGYGDLNGKEYWLVKNSWGHNFGEEGYIRMARNKGNHCGIASFPSYPEIVDHHHHHH
  • Zusammensetzung:
    Supplied as a 0.2 um filtered solution of 20mM MES, 150mM NaCl, 10% Glycerol, pH 5.5.
  • Validierte Anwendungen:
    SDS-PAGE
Spezifikationen
Zugehörige Informationen
Frequently Bought Together