Human recombinant CPA2 (from HEK293 cells)
Lieferant: Biorbyt
Zertifikate
Über diesen Artikel
Spezifikationen
- Konjugation:Unconjugated
- Protein/Peptid-Typ:Rekombinant
- Ursprung:HEK293 cells
- Spezies:Human
- Lagerbedingungen:Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Proteinsynonym:Carboxypeptidase A2
- UniProtKB:P48052 (Human)
- Protein/Peptid-Name:CPA2
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:LETFVGDQVLEIVPSNEEQIKNLLQLEAQEHLQLDFWKSPTTPGETAHVRVPFVNVQAVKVFLGS QGIAYSIMIEDVQVLLDKENEEMLFNRRRERSGNFNFGAYHTLEEISQEMDNLVAEHPGLVSKVN IGSSFENRPMNVLKFSTGGDKPAIWLDAGIHAREWVTQATALWTANKIVSDYGKDPSITSILDAL DIFLLPVTNPDGYVFSQTKNRMWRKTRSKVSGSLCVGVDPNRNWDAGFGGPGASSNPCSDSYHGP SANSEVEVKSIVDFIKSHGKVKAFITLHSYSQLLMFPYGYKCTKLDDFDELSEVAQKAAQSLRSL HGTKYKVGPICSVIYQASGGSIDWSYDYGIKYSFAFELRDTGRYGFLLPARQILPTAEETWLGLK AIMEHVRDHPYVDHHHHHH
- Zusammensetzung:Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl, 150mm NaCl, pH 7.5.
- Validierte Anwendungen:SDS-PAGE