Order Entry
Austria
ContactUsLinkComponent
Human recombinant IL6
Artikelnr. BIRBORB97797-20
Lieferant:  Biorbyt
CAS-Nummer:  
Human recombinant IL6
Artikelnr. BIRBORB97797-20
Lieferant:  Biorbyt
CAS-Nummer:  

Spezifikationen

  • VE:
    20 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Spezies:
    Human
  • Lagerbedingungen:
    Store at−20 °C.
  • Biologische Aktivität:
    ED50: Less than 0.1 ng/ml, corresponding to a specific activity of 1x107 IU/mg, determined by the dose dependent stimulation of murine 7TD1 cells.
  • Proteinsynonym:
    Interleukin BSF 2
  • UniProtKB:
    P05231
  • Protein/Peptid-Name:
    IL6
  • Purity:
    95% pure by SDS-PAGE and RP-HPLC
  • Solubility:
    It is recommended to reconstitute the lyophilized IL-6 inbidest water not less than 100 g/ml, which can then befurther diluted to other aqueous solutions. For long termstorage it is recommended to add a carrier protein (0.1%HSA or BSA).
  • Molecular weight:
    21 kDa
  • Zusammensetzung:
    Lyophilized. IL-6 is lyophilized from a concentrated PBSpH 7.4 solution.

Spezifikationen

Über diesen Artikel