ContactUsLinkComponent
Human IL13 Variant (from E. coli)
Human IL13 Variant (from E. coli)
Supplier:  Biorbyt
Certificates
Human IL13 Variant (from E. coli)
Supplier:  Biorbyt
Specifications
  • Ursprung:
    E. coli
  • Spezies:
    Human
  • Lagerbedingungen:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biologische Aktivität:
    The ED50 was determined by the dose dependent prolifiration of TF-1 cells and was found to be 1ng/ml, corresponding to specific activity of >1,000,000 units/mg. This analog has also been shown to exhibit increased in vivo activity compared to wild type IL-13.
  • Proteinsynonym:
    Allergic rhinitis
  • Protein/Peptid-Name:
    IL13 Variant
  • Purity:
    > 95% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Interleukin 13 Variant in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    SPGPVPPSTA LRELIEELVN ITQNQKAPLC NGSMVWSINL TAGMYCAALE SLINVSGCSA IEKTQRMLSG FCPHKVSAGQ FSSLHVRDTK IEVAQFVKDL LLHLKKLFRE GQFN.
  • Zusammensetzung:
    Lyophilized from 0.2 um filtered concentrated solution in 1xPBS, pH 7.2, containing 5% trehalose.
  • Validierte Anwendungen:
    SDS-PAGE , HPLC , FuncS
Specifications
Related Information
Frequently Bought Together